missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
Brand: Novus Biologicals NBP2-24683
This item is not returnable.
View return policy
Description
STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human, Mouse, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| STING/TMEM173 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| anti-STING, endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, human STING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, mouse STING, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, and dog, 94% homologous in mouse and cow, 88% homologous in rat, 83% homologous in opossum and zebrafish, and 72% homologous in Xenopus. Peptide blocking: The response seen in western blot is reduced by over 50% in the presence of immunizing peptide. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 1-5 ug/million cells, ELISA reported in scientific literature (PMID 34905508), Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 1:100 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 33745949) | |
| Q86WV6 | |
| TMEM173 | |
| Partial synthetic peptide made to an internal portion of human STING/TMEM173 (between amino acids 310-360) [UniProt Q86WV6] | |
| 0.1 mg | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Human, Mouse, Rat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction