missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
210.00 € - 634.00 €
Specifications
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
|---|---|
| Form | Purified |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Target Species | Human, Rat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18786863
|
Novus Biologicals
NBP2-24683SS |
0.025 mg |
210.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761574
|
Novus Biologicals
NBP2-24683 |
0.1 mg |
634.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human, Mouse, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Rabbit | |
| Human, Rat | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Purified | |
| RUO | |
| Partial synthetic peptide made to an internal portion of human STING/TMEM173 (between amino acids 310-360) [UniProt Q86WV6] |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title