missing translation for 'onlineSavingsMsg'
Learn More

Strontium Ranelate, Mikromol™

Product Code. 17964276
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
17964276 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 17964276

Brand: Mikromol MM3759.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Strontium Ranelate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Strontium Ranelate
CAS 135459-87-9
Analyte or Component Names Strontium Ranelate
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H6 N2 O8 S . 2 Sr
Certifications/Compliance ISO 17025
Synonym Bivalos, Distrontium renelate, Protelos, Protos, S 12911, S 12911-2 Strontium ranelate, 5-[bis(carboxymethyl)amino]-2-carboxy-4-cyano-3-thiopheneacetic acid strontium salt (1:2)
InChI Formula InChI=1S/C12H10N2O8S.2Sr/c13-2-6-5(1-7(15)16)10(12(21)22)23-11(6)14(3-8(17)18)4-9(19)20;;/h1,3-4H2,(H,15,16)(H,17,18)(H,19,20)(H,21,22);;/q;2*+2/p-4
SMILES [Sr+2].[Sr+2].[O-]C(=O)CN(CC(=O)[O-])c1sc(C(=O)[O-])c(CC(=O)[O-])c1C#N
IUPAC Name distrontium;5-[bis(carboxylatomethyl)amino]-3-(carboxylatomethyl)-4-cyanothiophene-2-carboxylate
Molecular Weight (g/mol) 513.49
Quantity 250 mg
Formula Weight 513.7957 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.