missing translation for 'onlineSavingsMsg'
Learn More

Suberoylanilide Hydroxamic Acid Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30428475
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428475 10 mg 10mg
30427798 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428475 Supplier Toronto Research Chemicals Supplier No. TRCS68871510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Suberoylanilide Hydroxamic Acid beta-D-Glucuronide Sodium Salt
Analyte or Component Names Suberoylanilide Hydroxamic Acid beta-D-Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C20H27N2NaO9
Synonym 1-O-[[1,8-Dioxo-8-(phenylamino)octyl]amino]-β-D-glucopyranuronic Acid Sodium Salt, Vorinostat glucuronide Sodium Salt
InChI Formula InChI=1S/C4H6N4O/c5-8-3-1-4(9)7-2-6-3/h1-2H,5H2,(H2,6,7,8,9)
Solution Type Neat
SMILES NNc1cc(O)ncn1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 462.43
Quantity 10 mg
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.