missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide Sodium Monohydrate, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Sulfacetamide Sodium Monohydrate
Specifications
Specifications
| Chemical Name or Material | Sulfacetamide sodium salt |
| CAS | 6209-17-2 |
| Analyte or Component Names | Sulfacetamide sodium salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H9 N2 Na O3 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Sulfacetamide Sodium Monohydrate, N-[(4-Aminophenyl)sulfonyl]acetamide sodium salt hydrate (1:1:1), N-[(4-Aminophenyl)sulfonyl]acetamide monosodium salt monohydrate, N-Sulfanilylacetamide monosodium salt monohydrate, N1-Acetylsulfanilamide sodium monohydrate, Sodium N-sulfanilylacetamide monohydrate, Sodium sulfacetamide monohydrate, Sulamyd sodium monohydrate, Sulfacetamide sodium monohydrate |
| SMILES | O.[Na+].CC(=O)[N-]S(=O)(=O)c1ccc(N)cc1 |
| Molecular Weight (g/mol) | 236.22 |
| Quantity | 500 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction