missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine EP Impurity A, Mikromol™

Product Code. 30050849
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050849 25 mg 25mg
30049324 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050849 Supplier Mikromol Supplier No. MM0623.030025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sulfadiazine EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrimidin-2-amine
CAS 109-12-6
Analyte or Component Names Pyrimidin-2-amine
Shipping Condition Room Temperature
Molecular Formula C4 H5 N3
Certifications/Compliance ISO 9001
Synonym Pyrimidin-2-amine, Sulfadiazine Imp. A (Pharmeuropa), Sulfadiazine Impurity A
InChI Formula InChI=1S/C4H5N3/c5-4-6-2-1-3-7-4/h1-3H,(H2,5,6,7)
SMILES Nc1ncccn1
IUPAC Name pyrimidin-2-amine
Molecular Weight (g/mol) 95.1
Quantity 25 mg
Formula Weight 95.0483 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.