missing translation for 'onlineSavingsMsg'
Learn More

Sulfaquinoxaline 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312838
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312838 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312838 Supplier Dr. Ehrenstorfer Supplier No. DREA16990000AL1000
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfaquinoxaline
CAS 59-40-5
Analyte or Component Names Sulfaquinoxaline
Shipping Condition +20°C
Molecular Formula C14 H12 N4 O2 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 4-Amino-N-2-quinoxalinyl-benzenesulfonamide, Sulfanilamide, N1-2-quinoxalinyl- (6CI,7CI,8CI), 2-(p-Sulfanilamido)quinoxaline, 2-Sulfanilamidobenzopyrazine, 2-p-Aminobenzenesulfonamidoquinoxaline, 2-p-Aminobenzenesulphonamidoquinoxaline, Avicocid, Compound 3-120, Italquina, Kokozigal, N1-(2-Quinoxalinyl)sulfanilamide, NSC 41805, SQ 40, SQX, Sulfa-Q 20, Sulfabenzpyrazine, Sulfaline, Sulfaquinoxaline, Sulphaquinoxaline, Sulquin, Ursokoxaline, Sulfachinoxalin
InChI Formula InChI=1S/C14H12N4O2S/c15-10-5-7-11(8-6-10)21(19,20)18-14-9-16-12-3-1-2-4-13(12)17-14/h1-9H,15H2,(H,17,18)
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2cnc3ccccc3n2
IUPAC Name 4-amino-N-quinoxalin-2-ylbenzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 300.34
Quantity 1 mL
Formula Weight 300.0681 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.