missing translation for 'onlineSavingsMsg'
Learn More

Sulpiride EP Impurity D, Mikromol™

Product Code. 30051797
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051797 25 mg 25mg
17901186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051797 Supplier Mikromol Supplier No. MM0051.040025
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sulpiride EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxy-5-sulfamoylbenzoic acid
CAS 22117-85-7
Analyte or Component Names 2-Methoxy-5-sulfamoylbenzoic acid
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O5 S
Certifications/Compliance ISO 17025
Synonym 2-Methoxy-5-sulphamoylbenzoic Acid, Sulpiride Imp. D (EP), Sulpiride Impurity D
InChI Formula InChI=1S/C8H9NO5S/c1-14-7-3-2-5(15(9,12)13)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)(H2,9,12,13)
SMILES COc1ccc(cc1C(=O)O)S(=O)(=O)N
IUPAC Name 2-methoxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 231.23
Quantity 25 mg
Formula Weight 231.0201 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.