missing translation for 'onlineSavingsMsg'
Learn More

SUPT5H Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30517185 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30517185 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30517185 Supplier Novus Biologicals Supplier No. NBP338598APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUPT5H Polyclonal antibody specifically detects SUPT5H in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT5H
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias DRB sensitivity-inducing factor 160 kDa subunit, DRB sensitivity-inducing factor large subunit, DSIF large subunit, DSIF p160, FLJ34157, SPT5hSPT5, SPT5HTat-cotransactivator 1 protein, suppressor of Ty (S.cerevisiae) 5 homolog, suppressor of Ty 5 homolog (S. cerevisiae), Tat-CT1, Tat-CT1 protein, transcription elongation factor SPT5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 125-425 of human SUPT5H (NP_003160.2).,, Sequence:, LQNLWRDQREEELGEYYMKKYAKSSVGETVYGGSDELSDDITQQQLLPGVKDPNLWTVKCKIGEERATAISLMRKFIAYQFTDTPLQIKSVVAPEHVKGYIYVEAYKQTHVKQAIEGVGNLRLGYWNQQMVPIKEMTDVLKVVKEVANLKPKSWVRLKRGIYKDDIAQVDYVEPSQNTISLKMIPRIDYDRIKARMSLKDWFAKRKKFKRPPQRLFDAEKIRSLGGDVASDGDFLIFEGNRYSRKGFLFKSFAMSAVITEGVKPTLSELEKFEDQPEGIDLEVVTESTGKEREHNFQPGDN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6829
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.