missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Neuraminidase Antibody - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Swine Influenza A H1N1 Neuraminidase Polyclonal antibody specifically detects Swine Influenza A H1N1 Neuraminidase in Virus samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Swine Influenza A H1N1 Neuraminidase |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1 ug/ml |
| Formulation | PBS with 0.02% Sodium Azide |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a synthetic peptide from the novel swine influenza Neuraminidase protein. This antibody is a cognate pair with product NBP2-41111. The immunogen is located within amino acids 50 - 100 of Swine H1N1 Neuraminidase. Amino Acid Squence: AAGQSVVSVK |
| Purification Method | Affinity Purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?