missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Antibody (SP15), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-97459-0.2ml
This item is not returnable.
View return policy
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Synaptophysin | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| SYP | |
| Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies. | |
| 0.2 mL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SP15 | |
| Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6855 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction