missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Antibody (SP15), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
162.00 € - 649.00 €
Specifications
| Antigen | Synaptophysin |
|---|---|
| Clone | SP15 |
| Dilution | Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038139
|
Novus Biologicals
NBP1-97459-0.025ML |
0.025 mL |
162.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014838
|
Novus Biologicals
NBP1-97459-0.05ML |
0.05 mL |
375.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775503
|
Novus Biologicals
NBP1-97459-0.2ml |
0.2 mL |
649.00 €
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Synaptophysin | |
| Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 6855 | |
| Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| SP15 | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human, Bovine | |
| Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
| SYP | |
| IgM | |
| Ammonium sulfate precipitation |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title