missing translation for 'onlineSavingsMsg'
Learn More

Syndecan-2/CD362 Antibody (305515), PE/Cy7, Novus Biologicals™

Product Code. 30031410 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30031410 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30031410 Supplier Novus Biologicals Supplier No. FAB2965PECY7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Syndecan-2/CD362 Monoclonal antibody specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan-2/CD362
Applications Flow Cytometry
Classification Monoclonal
Clone 305515
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD362 antigen, fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1syndecan proteoglycan 2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362, Glu19-Gly144, Accession # AAH49836.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 6383
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.