missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syrosingopine, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Syrosingopine |
| CAS | 84-36-6 |
| Analyte or Component Names | Syrosingopine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H42 N2 O11 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Yohimban-16-carboxylic acid, 18-[[4-[(ethoxycarbonyl)oxy]-3,5-dimethoxybenzoyl]oxy]-11,17-dimethoxy-, methyl ester, (3β,16β,17α,18β,20α)-, 3β,20α-Yohimban-16β-carboxylic acid, 18β-hydroxy-11,17α-dimethoxy-, methyl ester, 4-hydroxy-3,5-dimethoxybenzoate (ester) ethyl carbonate (ester) (8CI), Reserpic acid, methyl ester, ester with syringic acid, ethyl carbonate (6CI), Carbethoxysyringoyl methylreserpate, Isotense, Londomin, Methyl carbethoxysyringoyl reserpate, Methyl reserpate ester of syringic acid ethyl carbonate, NSC 77030, Raunova, Seniramin, Singoserp, Siringina, Su 3118, Syringic acid ethyl carbonate ester with methyl reserpate, Syringopine, Syrosingopin, Syrosingopine |
| InChI Formula | InChI=1S/C35H42N2O11/c1-7-46-35(40)48-31-26(42-3)12-18(13-27(31)43-4)33(38)47-28-14-19-17-37-11-10-22-21-9-8-20(41-2)15-24(21)36-30(22)25(37)16-23(19)29(32(28)44-5)34(39)45-6/h8-9,12-13,15,19,23,25,28-29,32,36H,7,10-11,14,16-17H2,1-6H3/t19-,23+,25-,28-,29+,32+/m1/s1 |
| Solution Type | Neat |
| SMILES | CCOC(=O)Oc1c(OC)cc(cc1OC)C(=O)O[C@@H]2C[C@@H]3CN4CCc5c([nH]c6cc(OC)ccc56)[C@H]4C[C@@H]3[C@@H]([C@H]2OC)C(=O)OC |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction