missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98919-100ul
This item is not returnable.
View return policy
Description
TACC3 Polyclonal antibody specifically detects TACC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TACC3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TACC3. | |
| 100 μg | |
| Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors | |
| 10460 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction