missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00 € - 326.00 €
Specifications
| Antigen | TACC3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624945
|
Novus Biologicals
NBP2-98919-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651766
|
Novus Biologicals
NBP2-98919-50ul |
50 μg |
241.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TACC3 Polyclonal antibody specifically detects TACC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TACC3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors | |
| 0.2 um filtered solution in PBS | |
| 10460 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TACC3. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title