missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACSTD2 / TROP2 (Epithelial Marker) Mouse, Clone: TACSTD2/2151, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4070-MSM1-P0
This item is not returnable.
View return policy
Specifications
| TACSTD2 / TROP2 (Epithelial Marker) | |
| Cell Surface | |
| TACSTD2/2151 | |
| Unconjugated | |
| Mouse | |
| 40kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 4070 | |
| IgG2b κ |
| Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human TACSTD2 protein (apprx. aa 31-274) | |
| HT29 cells. Breast or Colon Carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction