missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACSTD2 / TROP2 (Epithelial Marker) Mouse, Clone: TACSTD2/2151, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | TACSTD2 / TROP2 (Epithelial Marker) |
|---|---|
| Clone | TACSTD2/2151 |
| Concentration | 200μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15920408
|
EnquireBio
4070-MSM1-P0 |
20 μg | |||||||
|
15930408
|
EnquireBio
4070-MSM1-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| TACSTD2 / TROP2 (Epithelial Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human TACSTD2 protein (apprx. aa 31-274) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 40kDa |
| TACSTD2/2151 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 4070 | |
| IgG2b κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface | |
| HT29 cells. Breast or Colon Carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title