missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | TAF1L |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456981
|
Novus Biologicals
NBP2-30752-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18113723
|
Novus Biologicals
NBP2-30752 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF1L Polyclonal specifically detects TAF1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TAF1L | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8IZX4 | |
| 138474 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PKQPFMLQHASGEHKDGHGK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 210kDa, 210kDa-like, EC 2.7.11, EC 2.7.11.1, MGC134910, TAF(II)210, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF1-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2A2, TBP-associated factor 1-like, TBP-associated factor 210 kDa, TBP-associated factor RNA polymerase 1-like, Transcription initiation factor TFIID 210 kDa subunit, transcription initiation factor TFIID subunit 1-like | |
| TAF1L | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title