missing translation for 'onlineSavingsMsg'
Learn More

Tamibarotene

Product Code. 15446159
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446159 10 mg 10mg
15456159 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446159 Supplier Thermo Scientific Alfa Aesar Supplier No. J64028.MA
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

A retinoic acid receptor → (RAR→) agonist | CAS: 94497-51-5 | C22H25NO3 | 351.45 g/mol

Tamibarotene is a retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 ?M respectively).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tamibarotene is a retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 μM respectively).

Solubility
Soluble to 50 mM in DMSO

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94497-51-5
Molecular Formula C22H25NO3
Molecular Weight (g/mol) 351.45
MDL Number MFCD00866188
InChI Key MUTNCGKQJGXKEM-UHFFFAOYSA-N
Synonym tamibarotene, retinobenzoic acid, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl carbamoyl benzoic acid, amnolake, tamibaro, amnoid, unii-08v52gz3h9, 4-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl carbamoyl benzoic acid, chembl25202
PubChem CID 108143
ChEBI CHEBI:32181
IUPAC Name 4-[(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)carbamoyl]benzoic acid
SMILES CC1(C)CCC(C)(C)C2=CC(NC(=O)C3=CC=C(C=C3)C(O)=O)=CC=C12

Specifications

Color White
Quantity 10 mg
Merck Index 14,9047
Solubility Information Soluble to 50 mM in DMSO
Formula Weight 351.44
Physical Form Solid
Chemical Name or Material Tamibarotene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.