missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen Citrate, Mikromol™

Product Code. 18011750
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18011750 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18011750 Supplier Mikromol Supplier No. MM0053.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tamoxifen Citrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tamoxifen citrate
CAS 54965-24-1
Analyte or Component Names Tamoxifen citrate
Shipping Condition Room Temperature
Molecular Formula C26 H29 N O . C6 H8 O7
Certifications/Compliance ISO 17034
Synonym Tamoxifen Citrate, Ethanamine, 2-[4-[(1Z)-1,2-diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Ethanamine, 2-[4-(1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethyl-, (Z)-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Ethanamine, 2-[4-[(1Z)-1,2-diphenyl-1-butenyl]phenoxy]-N,N-dimethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1) (9CI), I.C.I.46474 citrate, ICI 46474, Istubal, Kessar, Noltam, Nolvadex, Nourytam, Soltamox, TMX, Tamofen, Tamoplex, Tamox-Puren, Tomaxasta, Valodex, Z-Tamoxifen citrate, Zemide
InChI Formula InChI=1S/C26H29NO.C6H8O7/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,4,19-20H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;
SMILES CC\C(=C(/c1ccccc1)\c2ccc(OCCN(C)C)cc2)\c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 563.64
Quantity 100 mg
Formula Weight 563.2519 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.