missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54723-25ul
This item is not returnable.
View return policy
Description
TCEAL5 Polyclonal specifically detects TCEAL5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TCEAL5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| TCEA-like protein 5, transcription elongation factor A (SII)-like 5, transcription elongation factor A protein-like 5, Transcription elongation factor S-II protein-like 5 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 340543 | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TCEAL5 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:PKDSQEDLQERHLSSEEMMRECGDVSRAQEELRK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction