missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCERG1L Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05852-50ul
This item is not returnable.
View return policy
Description
TCERG1L Monoclonal antibody specifically detects TCERG1L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TCERG1L | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 256536 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 05 | |
| Immunohistochemistry-Paraffin 1:250-1:1000 | |
| 5730476P14Rik, FLJ38950, MGC126584, transcription elongation regulator 1-like, transcription elongation regulator 1-like protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human TCERG1L. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction