missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCERG1L Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | TCERG1L |
|---|---|
| Clone | 05 |
| Dilution | Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690058
|
Novus Biologicals
NBP3-05852-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659336
|
Novus Biologicals
NBP3-05852-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCERG1L Monoclonal antibody specifically detects TCERG1L in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TCERG1L | |
| Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 256536 | |
| IgG1 | |
| Protein A purified |
| 05 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| 5730476P14Rik, FLJ38950, MGC126584, transcription elongation regulator 1-like, transcription elongation regulator 1-like protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human TCERG1L. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title