missing translation for 'onlineSavingsMsg'
Learn More

TCP1-delta Antibody, DyLight 755, Novus Biologicals™

Product Code. 18678369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18678369 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18678369 Supplier Novus Biologicals Supplier No. NBP241360IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCP1-delta Polyclonal antibody specifically detects TCP1-delta in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP1-delta
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CCTD, CCT-DELTA, chaperonin containing t-complex polypeptide 1, delta subunit, chaperonin containing TCP1, subunit 4 (delta), MGC126165, SRBMGC126164, Stimulator of TAR RNA-binding, T-complex protein 1 subunit delta, TCP-1-delta
Host Species Rabbit
Immunogen The epitope is recognized to a region between residue 1 to 50 of human Chaperonin Containing Tcp1, Subunit 4 (Delta).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10575
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.