missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDAG8/GPR65 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21356-25ul
This item is not returnable.
View return policy
Description
TDAG8/GPR65 Polyclonal antibody specifically detects TDAG8/GPR65 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TDAG8/GPR65 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| G protein-coupled receptor 65, G-protein coupled receptor 65, psychosine receptor, T-cell death-associated gene 8 protein, TDAG8hTDAG8 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LLIRCILEHAVNFEDHSNSGKRTYTMYRI | |
| 25 μL | |
| Apoptosis, GPCR | |
| 8477 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction