missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDAG8/GPR65 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
401.00 € - 653.00 €
Specifications
| Antigen | TDAG8/GPR65 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683654
|
Novus Biologicals
NBP3-21356-25ul |
25 μL |
401.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619853
|
Novus Biologicals
NBP3-21356-100ul |
100 μL |
653.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TDAG8/GPR65 Polyclonal antibody specifically detects TDAG8/GPR65 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TDAG8/GPR65 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, GPCR | |
| PBS, pH 7.2, 40% glycerol | |
| 8477 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor 65, G-protein coupled receptor 65, psychosine receptor, T-cell death-associated gene 8 protein, TDAG8hTDAG8 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LLIRCILEHAVNFEDHSNSGKRTYTMYRI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title