missing translation for 'onlineSavingsMsg'
Learn More

TDRD3 Antibody (RAB-C419), Alexa Fluor™ 405 - Chimeric, R&D Systems™

Product Code. 18611980 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611980 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611980 Supplier Novus Biologicals Supplier No. NBP320149AF405
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

TDRD3 Monoclonal specifically detects TDRD3 in Human samples. It is validated for ChIP Assay∣ELISA∣Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Immunoprecipitation∣Mass Spectrometry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD3
Applications ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, Western Blot
Classification Monoclonal
Clone RAB-C419
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias FLJ21007, tudor domain containing 3, tudor domain-containing protein 3
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing TDRD3 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81550
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.