missing translation for 'onlineSavingsMsg'
Learn More

Temozolomide, Mikromol™

Product Code. 18010570
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18010570 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18010570 Supplier Mikromol Supplier No. MM1480.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Temozolomide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Temozolomide
CAS 85622-93-1
Analyte or Component Names Temozolomide
Shipping Condition Room Temperature
Molecular Formula C6 H6 N6 O2
Certifications/Compliance ISO 17034
Synonym Temozolomide, 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5]tetrazinecarboxamide, 3-Methyl-8-carbamoylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one, 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one, CCRG 81045, M and B 39831, MB 39831, Methazolastone, NSC 362856, Sch 52365, Temodal, Temodar, Imidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide, 3,4-dihydro-3-methyl-4-oxo-
InChI Formula InChI=1S/C6H6N6O2/c1-11-6(14)12-2-8-3(4(7)13)5(12)9-10-11/h2H,1H3,(H2,7,13)
SMILES CN1N=Nc2c(ncn2C1=O)C(=O)N
IUPAC Name 3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
Molecular Weight (g/mol) 194.15
Quantity 250 mg
Formula Weight 194.0552 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.