missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-[(1S,2R)-1-Benzyl-2-hydroxy-3-[(2-methylpropyl)amino]propyl]carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
tert-Butyl N-[(1S,2R)-1-Benzyl-2-hydroxy-3-[(2-methylpropyl)amino]propyl]carbamate
Specifications
Specifications
| Chemical Name or Material | tert-Butyl [(1S,2R)-1-Benzyl-2-hydroxy-3-(isobutylamino)prop |
| CAS | 160232-08-6 |
| Analyte or Component Names | tert-Butyl [(1S,2R)-1-Benzyl-2-hydroxy-3-(isobutylamino)prop |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H32 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Carbamic acid, N-[(1S,2R)-2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, Carbamic acid, [(1S,2R)-2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester (9CI), Carbamic acid, [2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, [R-(R*,S*)]-, tert-Butyl [(1S,2R)-1-benzyl-2-hydroxy-3-(isobutylamino)propyl]carbamate |
| InChI Formula | InChI=1S/C19H32N2O3/c1-14(2)12-20-13-17(22)16(11-15-9-7-6-8-10-15)21-18(23)24-19(3,4)5/h6-10,14,16-17,20,22H,11-13H2,1-5H3,(H,21,23)/t16-,17+/m0/s1 |
| SMILES | CC(C)CNC[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-[(2S,3R)-3-hydroxy-4-(2-methylpropylamino)-1-phenylbutan-2-yl]carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction