missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylmagnesium chloride, 1.7M solution in THF, AcroSeal™
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Linear Formula | (CH3)3CMgCl |
| Fieser | 04,63; 12,171 |
| Density | 0.9310g/mL |
| IUPAC Name | magnesium;2-methylpropane;chloride |
| Formula Weight | 116.87 |
| Percent Purity | 19 to 21% (as BUMC) |
| Packaging | AcroSeal™ Glass bottle |
| Specific Gravity | 0.931 |
| Quantity | 800 mL |
| Show More |
Safety and Handling
Product Identifier
- tert-Butylmagnesium chloride, 1.7M solution in THF
- Danger
- Flammable liquid Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: tert-Butylmagnesiumchloride, Tetrahydrofuran
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?