missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Benzoate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Testosterone Benzoate |
| CAS | 2088-71-3 |
| Analyte or Component Names | Testosterone Benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H32 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Testosterone benzoate, Testosterone, benzoate (7CI,8CI), NSC 524278, Testosterone 17-benzoate, Androst-4-en-3-one, 17-(benzoyloxy)-, (17beta)-, (17beta)-17-(Benzoyloxy)androst-4-en-3-one, 17beta-Benzoyloxyandrost-4-en-3-one, 17beta-Hydroxyandrost-4-en-3-one benzoate, Androst-4-en-17beta-ol-3-one benzoate |
| InChI Formula | InChI=1S/C26H32O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h3-7,16,20-23H,8-15H2,1-2H3/t20-,21-,22-,23-,25-,26-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)c5ccccc5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?