missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Propionate, Mikromol™

Product Code. 18051020
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18051020 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18051020 Supplier Mikromol Supplier No. MM0897.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Testosterone Propionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone propionate
CAS 57-85-2
Analyte or Component Names Testosterone propionate
Shipping Condition Room Temperature
Molecular Formula C22 H32 O3
Certifications/Compliance ISO 17025
Synonym Testosterone Isocaproate Imp. C (EP), Testosterone Propionate, Androst-4-en-3-one, 17-(1-oxopropoxy)-, (17beta)-, Testosterone, propionate (8CI), (17beta)-17-(1-Oxopropoxy)androst-4-en-3-one, 17beta-(Propionyloxy)androst-4-en-3-one, 17beta-Hydroxy-4-androsten-3-one 17-propionate, 17beta-Hydroxyandrost-4-en-3-one propionate, 17beta-Hydroxyandrost-4-en-3-one-17beta-propionate, Agovirin, Androlon, Androtest P, Androteston, Anertan, Aquaviron, Bio-testiculina, Enarmon, Enarmon-oil, Homandren, Hormoteston, Masenate, NRB 03689, NSC 9166, Nasdol, Okasa-Mascul, Orchiol, Orchisterone, Orchistin, Oreton, Pantestin, Primotestone, Propiokan, Recthormone testosterone, Solvotest, Sterandryl, Synandrol, Synerone, TP, Telipex, Testaform, Testex, Testodet, Testodrin, Testogen, Testolets, Testonique, Testormol, Testosid, Testosteron propionate, Testosterone-17-propionate, Testosterone-17beta propionate, Testoviron, Testoviron (ampule), Testoxyl, Testrex, Textes Depot, Tostrin, Uniteston, Virormone, neo-Hombreol, Testosterone Isocaproate Impurity C
InChI Formula InChI=1S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1
SMILES CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 344.49
Quantity 100 mg
Formula Weight 344.2351 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.