missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TET1 Polyclonal antibody specifically detects TET1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | TET1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TET1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?