missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine, LoGiCal

Product Code. 30460082
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30460082 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30460082

Brand: LoGiCal LGCFOR3473.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetracaine
CAS 94-24-6
Analyte or Component Names Tetracaine
Shipping Condition Room Temperature
Molecular Formula C15 H24 N2 O2
Certifications/Compliance ISO 9001
Synonym Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester (8CI), 2-Dimethylaminoethyl p-butylaminobenzoate, Amethocaine, Ametop, Anetain, Butylocaine, Contralgin, Fissucain, Intercain, Laudocaine, Medicaine, Medihaler-Tetracaine, Meethobalm, Metraspray, Mucaesthin, Pontocaine, Rexocaine, Tetracaine, Uromucaesthin, beta-Dimethylaminoethyl p-N-butylaminobenzoate, beta-Dimethylaminoethyl p-butylaminobenzoate
InChI Formula InChI=1S/C15H24N2O2/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3/h6-9,16H,4-5,10-12H2,1-3H3
Solution Type Neat
SMILES CCCCNc1ccc(cc1)C(=O)OCCN(C)C
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 264.36
Quantity 10 mg
Formula Weight 264.1838 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.