missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine, Mikromol™

Product Code. 18043850
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
18043850 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 18043850

Brand: Mikromol MM3473.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tetracaine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetracaine
CAS 94-24-6
Analyte or Component Names Tetracaine
Shipping Condition Room Temperature
Molecular Formula C15 H24 N2 O2
Certifications/Compliance ISO 17025
Synonym Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester (8CI), 2-Dimethylaminoethyl p-butylaminobenzoate, Amethocaine, Ametop, Anetain, Butylocaine, Contralgin, Fissucain, Intercain, Laudocaine, Medicaine, Medihaler-Tetracaine, Meethobalm, Metraspray, Mucaesthin, Pontocaine, Rexocaine, Tetracaine, Uromucaesthin, beta-Dimethylaminoethyl p-N-butylaminobenzoate, beta-Dimethylaminoethyl p-butylaminobenzoate
InChI Formula InChI=1S/C15H24N2O2/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3/h6-9,16H,4-5,10-12H2,1-3H3
SMILES CCCCNc1ccc(cc1)C(=O)OCCN(C)C
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate
Molecular Weight (g/mol) 264.36
Quantity 250 mg
Formula Weight 264.1838 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.