missing translation for 'onlineSavingsMsg'
Learn More

TGF-alpha, Rabbit anti-Human, Clone: tAb2, Novus Biologicals™

Product Code. p-7182737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.20mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329560 0.025 mg 25µg
18397990 0.2 mg 0.20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18329560 Supplier Novus Biologicals Supplier No. NBP2810820.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for Flow Cytometry, ELISA, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-alpha
Applications ELISA, Dot Blot
Classification Monoclonal
Clone tAb2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Dot Blot
Formulation PBS
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Gene Symbols TGFA
Host Species Rabbit
Immunogen Biologically active hemocyanin-coupled human TGF-alpha.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Test Specificity tAb2 binds the N-terminal sequence of TGF-alpha, VVSHFND (with residues 4 to 7 being the minimal binding epitope).
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.