missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN38 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NBP1-03495
This item is not returnable.
View return policy
Description
TGN38 Polyclonal specifically detects TGN38 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Spezifikation
| TGN38 | |
| Polyclonal | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
| Q62314 | |
| TGOLN2 | |
| A synthetic peptide from mouse TGN38 conjugated to blue carrier protein was used as the antigen. | |
| 0.5 mg | |
| Golgi Apparatus Markers, Trans golgi Network Markers | |
| 10618 | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Contains 0.02% benzalkonium chloride. with No Preservative | |
| TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| TGN38 (or TGN46, TGN48, TGN51 in human). | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur