missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN38 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
260.00 € - 557.00 €
Specifications
| Target Species | Human, Mouse |
|---|---|
| Isotype | IgG |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18748333
|
Novus Biologicals
NBP1-03495SS |
260.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18202760
|
Novus Biologicals
NBP1-03495 |
0.5 mg |
557.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGN38 Polyclonal specifically detects TGN38 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Human, Mouse | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title