missing translation for 'onlineSavingsMsg'
Learn More

Theobromine 0.1 mg/ml in Methanol, LoGiCal

Product Code. 30462499
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462499 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462499 Supplier LoGiCal Supplier No. LGCAMP0577.0002

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Theobromine
CAS 83-67-0
Analyte or Component Names Theobromine
Shipping Condition Room Temperature
Molecular Formula C7 H8 N4 O2
Certifications/Compliance ISO 17034
Synonym Caffeine Imp. D (EP), Pentoxifylline Imp. A (EP), Theophylline-Ethylenediamine Anhydrous Imp. G (EP), Theobromine, Theophylline-Ethylenediamine Imp. G (EP), Caffeine Impurity D, Pentoxifylline Impurity A, Theophylline-Ethylenediamine Impurity G
InChI Formula InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)
Solution Type Single Solution
SMILES CN1C(=O)NC(=O)c2c1ncn2C
IUPAC Name 3,7-dimethylpurine-2,6-dione
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 180.16
Quantity 1 mL
Formula Weight 180.0647 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.