missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Thermal Transfer Printable Labels, B-424

Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of general purpose paper (B-424).

67.84 € - 279.80 €

Specifications

Description Recommended ribbon series: R4300; R4500; R6100
Adhesive Type Permanent Rubber
Coating None
Color White
Finish Matte
View More Specs

Products 4
Product Code Brand Size (Metric) Price Quantity & Availability  
Product Code Brand Size (Metric) Price Quantity & Availability  
11503724
View Documents Promotion Details
Brady™
030132
50.8 x 25.4 mm (W x H)
ONLINE EXCLUSIVE PRICE
70.50 €
67.84 € / 1 rolls
Save 2.66 €
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 30-10-2026
Add to basket
 
11533724
View Documents Promotion Details
Brady™
030135
76.2 x 25.4 mm (W x H)
ONLINE EXCLUSIVE PRICE
107.00 €
102.06 € / 1 rolls
Save 4.94 €
4% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
11593724
View Documents
Brady™
030141
W x H: 76.2 mm x 191.44m
ONLINE EXCLUSIVE PRICE
136.00 €
130.46 € / 1 rolls
Save 5.54 €
4% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
13019242
View Documents
Brady™
030153
101.6 x 152.4 mm (W x H)
ONLINE EXCLUSIVE PRICE
291.00 €
279.80 € / 1 rolls
Save 11.20 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
Description

Description

  • For use with thermal transfer printers
  • Made of contrast and smear resistant paper
Specifications

Specifications

Recommended ribbon series: R4300; R4500; R6100
None
Matte
Paper (B-424)
General and industrial identification
0.0035 in.
Permanent Rubber
White
Roll
Non-sterile
Rectangle
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.