missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Thiocarbamate, TRC

Product Code. 30456857
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456857 100 mg 100mg
30455446 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456857 Supplier Toronto Research Chemicals Supplier No. TRCT344093100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamine Thiocarbamate
Analyte or Component Names Thiamine Thiocarbamate
Shipping Condition Room Temperature
Molecular Formula C14H20N4O3S2
Synonym [(4-Amino-2-methyl-5-pyrimidinyl)methyl]carbamodithioic Acid 1-[2-(Acetyloxy)ethyl]-2-oxopropyl Ester + 2-(3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-4-hydroxy-4-methyl-2-thioxothiazolidin-5-yl)ethyl acetate
Solution Type Neat
SMILES -NC1=C(CN2C(SC(CCOC(C)=O)C2(O)C)=S)C=NC(C)=N1.NC3=C(CNC(SC(C(C)=O)CCOC(C)=O)=S)C=NC(C)=N3
Industry Type Pharmaceutical
Molecular Weight (g/mol) 356.46
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.