missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzoic acid, 90%

Product Code. 10381334 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10381334

Brand: Thermo Scientific Acros 138685000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 98-91-9 | C7H6OS | 138.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-91-9
MDL Number MFCD00004852
InChI Key UIJGNTRUPZPVNG-UHFFFAOYSA-N
Synonym thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n
PubChem CID 7414
IUPAC Name benzenecarbothioic S-acid
SMILES C1=CC=C(C=C1)C(=O)S

Specifications

Melting Point 15.0°C to 18.0°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 122.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C7H6OS
Linear Formula C6H5COSH
Refractive Index 1.6020 to 1.6080
Quantity 500 g
Beilstein 09, 419
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Molecular Weight (g/mol) 138.18
Formula Weight 138.18
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Thiobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Thiobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.