missing translation for 'onlineSavingsMsg'
Learn More

Thioctic Acid, Mikromol™

Product Code. 17984306
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17984306 Supplier Mikromol Supplier No. MM0127.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Thioctic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-alpha-Lipoic Acid
CAS 1077-28-7
Analyte or Component Names DL-alpha-Lipoic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H14 O2 S2
Certifications/Compliance ISO 17034
Synonym Thioctic Acid, 1,2-Dithiolane-3-pentanoic acid, (RS)-Lipoic acid, 1,2-Dithiolane-3-valeric acid, 5-(1,2-Dithiolan-3-yl)pentanoic acid, 5-(1,2-Dithiolan-3-yl)valeric acid, 6,8-Thioctic acid, 6-Thioctic acid, Alipure, Biletan, DL-6,8-Thioctic acid, DL-6-Thioctic acid, DL-Thioctic acid, Espa-lipon, Liposan, Lipothion, NSC 628502, NSC 90788, Protogen A, Pyruvate oxidation factor, Thioctacid, Thioctic acid, Thioctsan, Thiotacid, Tioctacid, Tioctidasi, Tioctidasi acetate replacing factor, dl-Lipoic acid, dl-Thioctic acid, (+/-)-1,2-Dithiolane-3-pentanoic acid, (+/-)-1,2-Dithiolane-3-valeric acid, (+/-)-alpha-Lipoic acid, (+/-)-Lipoic acid, (+/-)-Thioctic acid, (RS)-alpha-Lipoic acid, alpha-Liponic acid, DL-alpha-Lipoic acid
InChI Formula InChI=1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)
SMILES OC(=O)CCCCC1CCSS1
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
Molecular Weight (g/mol) 206.33
Quantity 500 mg
Formula Weight 206.0435 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.