missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxylic Acid, Mikromol™

Product Code. 30052481
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052481 100 mg 100mg
30051863 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052481 Supplier Mikromol Supplier No. MM1081.04
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Thiophene-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiophene-3-carboxylic Acid
CAS 88-13-1
Analyte or Component Names Thiophene-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C5 H4 O2 S
Certifications/Compliance ISO 9001
Synonym Thiophene-3-carboxylic Acid, 3-Thiophenecarboxylic acid, 3-Carboxythiophene, 3-Thenoic acid, 3-Thienylcarboxylic acid, NSC 66314, beta-Thiophenecarboxylic acid, beta-Thiophenic acid
InChI Formula InChI=1S/C5H4O2S/c6-5(7)4-1-2-8-3-4/h1-3H,(H,6,7)
SMILES OC(=O)c1ccsc1
IUPAC Name thiophene-3-carboxylic acid
Molecular Weight (g/mol) 128.15
Quantity 100 mg
Formula Weight 127.9932 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.