missing translation for 'onlineSavingsMsg'
Learn More

Thomsen-Friedenreich Antigen Antibody (A68-B/A11), Novus Biologicals™

Product Code. 18185739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18185739 0.02 mg 0.02mg
18176209 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 18185739 Proveedor Novus Biologicals N.º de proveedor NBP2452760.02MG

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence

Thomsen-Friedenreich Antigen Monoclonal specifically detects Thomsen-Friedenreich Antigen in Human, Mouse, Rat samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen Thomsen-Friedenreich Antigen
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone A68-B/A11
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Host Species Mouse
Immunogen Neuraminidase-treated human red blood cells
Purification Method Protein A or G purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a disaccharide epitope, Gal1-3GalNAc, of Thomsen-Friedenreich (TF) antigen. It is specific for both anomeric forms of the disaccharide (TF and TF, including related structures on the glycolipid) and shows no cross-reactivity with sialylated glycophorin. The Thomsen-Friedenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered as a pan-carcinoma marker. This MAb is capable to agglutinate desialylated red blood cells. During metastasis, the ability of malignant cells to form multicellular aggregates via homotypic or heterotypic aggregation and their adhesion to the endothelium are critical. The tumor-associated carbohydrate Thomsen-Friedenreich antigen (Gal-GalNAc) is involved in tumor cell adhesion and tissue invasion. It also causes an immune response, and overexpression of the antigen causes cancer cells to be more sensitive to natural killer cell lysis. The Thomsen-Friedenreich antigen is suppressed in normal healthy cells and represents one of the few chemically well-defined antigens associated with tumor malignancy. The presence of the Thomsen-Friedenreich antigen on the surface of cancer cells may result from a divergence from the normal pathway for O-linked glycosylation in these cells, most likely caused by inappropriate localization of the enzymes involved in synthesis of the disaccharide.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgM κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.