missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24477
This item is not returnable.
View return policy
Description
THRAP3 Polyclonal specifically detects THRAP3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| THRAP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS + 0.05% BSA with 0.05% Sodium Azide | |
| FLJ22082, MGC133082, MGC133083, thyroid hormone receptor associated protein 3, Thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit, Trap150, TRAP150thyroid hormone receptor-associated protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), horse, cow, dog, mouse, monkeyand chimpanzee, 93% homologous in chicken, 78% homologous in xenopus and 73% homologous in rat and opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q9Y2W1 | |
| THRAP3 | |
| A portion of amino acids 930-955 of human Thyroid hormone receptor associated protein 3 was used as the immunogen | |
| 0.1 mg | |
| Signal Transduction, Transcription Factors and Regulators | |
| 9967 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction