missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 351.00 €
Specifications
| Antigen | THRAP3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18160012
|
Novus Biologicals
NBP2-24477SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700614
|
Novus Biologicals
NBP2-24477 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
THRAP3 Polyclonal specifically detects THRAP3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| THRAP3 | |
| Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction, Transcription Factors and Regulators | |
| PBS + 0.05% BSA with 0.05% Sodium Azide | |
| FLJ22082, MGC133082, MGC133083, thyroid hormone receptor associated protein 3, Thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit, Trap150, TRAP150thyroid hormone receptor-associated protein 3 | |
| THRAP3 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), horse, cow, dog, mouse, monkeyand chimpanzee, 93% homologous in chicken, 78% homologous in xenopus and 73% homologous in rat and opossum. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Primate | |
| Q9Y2W1 | |
| 9967 | |
| A portion of amino acids 930-955 of human Thyroid hormone receptor associated protein 3 was used as the immunogen | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title