missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thromboxane A2 R/TBXA2R Antibody (SR1893), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22509-100ul
This item is not returnable.
View return policy
Description
Thromboxane A2 R/TBXA2R Monoclonal antibody specifically detects Thromboxane A2 R/TBXA2R in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Thromboxane A2 R/TBXA2R | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1893 | |
| Western Blot 1:500-1:2000 | |
| Prostanoid TP receptor, thromboxane A2 receptor, TXA2-R | |
| A synthesized peptide derived from human Thromboxane A2 R/TBXA2R (Uniprot #: P21731) | |
| 100 μg | |
| GPCR | |
| 6915 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction