missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thromboxane A2 R/TBXA2R Antibody (SR1893), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Thromboxane A2 R/TBXA2R |
|---|---|
| Clone | SR1893 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698584
|
Novus Biologicals
NBP3-22509-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689734
|
Novus Biologicals
NBP3-22509-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thromboxane A2 R/TBXA2R Monoclonal antibody specifically detects Thromboxane A2 R/TBXA2R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Thromboxane A2 R/TBXA2R | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Prostanoid TP receptor, thromboxane A2 receptor, TXA2-R | |
| A synthesized peptide derived from human Thromboxane A2 R/TBXA2R (Uniprot #: P21731) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1893 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6915 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title